Ac-Phe-Gly-Pna

Supplier: BACHEM AMERICAS INC

4005053.0050 4005053.0250
L-1060.0050BAEA 309.66 CAD
L-1060.0050BA L-1060.0250BA
Ac-Phe-Gly-Pna
Ac-Phe-Gly-Pna

Sequence: Ac-Phe-Gly-pNA


Chromogenic papain substrate.

Formula: C₁₉H₂₀N₄O₅
Storage Temperature: Freezer
MDL Number: MFCD00155538
CAS Number: 34336-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR