Ac-Lys-pNA

Supplier: BACHEM AMERICAS INC

4004444.0050 4004444.0250
L-1045.0050BAEA 325.67 CAD
L-1045.0050BA L-1045.0250BA
Ac-Lys-pNA
Ac-Lys-pNA

Sequence: Ac-Lys-pNA


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.


Chromogenic substrate for gingipain K.

Formula: C₁₄H₂₁ClN₄O₄
MW: 344.796 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237501
CAS Number: 50931-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR