Ac-Arg-Pna·HCl

Supplier: BACHEM AMERICAS INC

4004007.0250 4004007.1000
L-1025.0250BAEA 688.11 CAD
L-1025.0250BA L-1025.1000BA
Ac-Arg-Pna·HCl
Ac-Arg-Pna·HCl

Sequence: Ac-Arg-pNA · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Synonym(s): L-AAPA

Formula: C₁₄H₂₁ClN₆O₄
Storage Temperature: Freezer
MDL Number: MFCD00155536
CAS Number: 40127-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR