Pyr-βNA

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-N-(Naphthalen-2-yl)-5-oxopyrrolidine-2-carboxamide, L-Pyroglutamic acid beta-naphthylamide

4003907.0250 4003907.1000 4003907.5000
K-1480.0250BAEA 180.91 CAD
K-1480.0250BA K-1480.1000BA K-1480.5000BA
Pyr-βNA
Pyr-βNA

Sequence: Pyr-βNA


Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.


Substrate for the assay of pyroglutamyl peptidase I.

Formula: C₁₅H₁₄N₂O₂
MW: 254.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055911
CAS Number: 22155-91-5
EINECS: 244-809-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR