L-Proline-β-naphthylamide hydrochloride

Supplier: BACHEM AMERICAS INC

4000630.0001 4000630.0005
K-1465.0001BAEA 180.83 CAD
K-1465.0001BA K-1465.0005BA
L-Proline-β-naphthylamide hydrochloride
L-Proline-beta-naphthylamide hydrochloride

Sequence: H-Pro-βNA · HCl


Substrate for the determination of proline arylamidase in human sera by the endpoint-method. It is also readily hydrolyzed by prolyl aminopeptidase from Bacillus megaterium.

Formula: C₁₅H₁₇ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00038937
CAS Number: 97216-16-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR