Phe-Arg β-naphthylamide dihydrochloride

Supplier: BACHEM AMERICAS INC

4000885.0250 4000885.1000
K-1450.0250BAEA 424.15 CAD
K-1450.0250BA K-1450.1000BA
Phe-Arg β-naphthylamide dihydrochloride
Phe-Arg beta-naphthylamide dihydrochloride

Sequence: H-Phe-Arg-βNA · 2 HCl


Antimicrobial peptides are produced by plants and most organisms throughout the animal kingdom including humans. Antimicrobial peptides protect against a broad range of infectious agents, as bacteria, fungi, and viruses. The amphibian skin is an especially rich source of antimicrobial peptides. See also the product families: Hepcidins LL-37 and Fragments Tuftsin and Analogs (subfamily).


Synonym(s): MC-207110#PAbN

Formula: C₂₅H₃₂Cl₂N₆O₂
Storage Temperature: Freezer
MDL Number: MFCD00058046
CAS Number: 100929-99-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR