L-Leucine-β-naphthylamide hydrochloride

Supplier: BACHEM AMERICAS INC

4000641.0001 4000641.0005
K-1385.0001BAEA 180.91 CAD
K-1385.0001BA K-1385.0005BA
L-Leucine-β-naphthylamide hydrochloride
L-Leucine-2-naphthylamide hydrochloride

Sequence: H-Leu-βNA · HCl


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).
Commonly used protease inhibitors as bestatin and the E-64s are derived from leucine. See also H-Leu-NH₂ (E-2650) and Z-Leu-ONp (C-2120).


(see also H-Leu-βNA (K-1380))

Formula: C₁₆H₂₁ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00034972
CAS Number: 893-36-7
EINECS: 212-970-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR