H-Leu-βNA

Supplier: BACHEM AMERICAS INC

4001935.0001 4001935.0005
K-1380.0001BAEA 180.91 CAD
K-1380.0001BA K-1380.0005BA
H-Leu-βNA
H-Leu-βNA

Sequence: H-Leu-βNA


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).
Commonly used protease inhibitors as bestatin and the E-64s are derived from leucine. See also H-Leu-NH₂ (E-2650) and Z-Leu-ONp (C-2120).


Substrate for assays of aminopeptidase M and leucine aminopeptidase. Its use for histochemical purposes has been described.

Formula: C₁₆H₂₀N₂O
MW: 256.35 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00055914
CAS Number: 732-85-4
EINECS: 211-990-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR