Glycylphenylalanine-2-naphthylamide
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Gly-Phe-βNA
4001208.0250
4001208.1000
K-1325.0250BAEA
413.58
CAD
K-1325.0250BA
K-1325.1000BA
Glycylphenylalanine-2-naphthylamide
Glycylphenylalanine-2-naphthylamide
Sequence: H-Gly-Phe-βNA
Synonym(s): GPN
GF-βNA (GPN), standard substrate for assays of dipeptidyl aminopeptidase I (cathepsin C). GPN causes osmotic permeabilization of lysosomes due to intralysosomal hydrolysis by cathepsin C.
Formula:
C₂₁H₂₁N₃O₂ MW: 347.418 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00021597 CAS Number: 21438-66-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...