H-Gly-Asp-Asp-Asp-Asp-Lys-betaNA
Supplier: BACHEM AMERICAS INC
4001598.0005 4001598.0025
K-1310.0005BAEA 217.06 CAD
K-1310.0005BA K-1310.0025BA
H-Gly-Asp-Asp-Asp-Asp-Lys-betaNA
Proteins and Peptides
GDDDDK-βNA, specific substrate for the determination of enteropeptidase (enterokinase), also used in histochemistry.