Glycine-β-naphthylamide hydrochloride

Supplier: BACHEM AMERICAS INC

4001855.0001 4001855.0005
K-1295.0001BAEA 413.58 CAD
K-1295.0001BA K-1295.0005BA
Glycine-β-naphthylamide hydrochloride
Glycine-beta-naphthylamide hydrochloride

Sequence: H-Gly-βNA · HCl


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.
See also Boc-Gly-ONp (A-1740) and Z-Gly-ONp (M-1250).

Formula: C₁₂H₁₃ClN₂O
Storage Temperature: Ambient
MDL Number: MFCD00051282
CAS Number: 1208-12-4
EINECS: 214-899-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR