H-Glu(βNA)-OH

Supplier: BACHEM AMERICAS INC

4003731.0250 4003731.1000
K-1270.0250BAEA 309.81 CAD
K-1270.0250BA K-1270.1000BA
H-Glu(βNA)-OH
H-Glu(βNA)-OH

Sequence: H-Glu(βNA)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
See also the glutamic acid γ-diazomethyl ketones L-DON (F-1510) and D-DON (F-2185).


Specific and sensitive substrate for the determination of serum γ-glutamyl transferase activity.

Formula: C₁₅H₁₆N₂O₃
MW: 272.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00037743
CAS Number: 14525-44-1
EINECS: 238-542-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR