H-Glu-βNA

Supplier: BACHEM AMERICAS INC

4001467.0001 4001467.0005
K-1265.0001BAEA 258.14 CAD
K-1265.0001BA K-1265.0005BA
H-Glu-βNA
H-Glu-βNA

Sequence: H-Glu-βNA


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
See also the glutamic acid γ-diazomethyl ketones L-DON (F-1510) and D-DON (F-2185).


Specific substrate for aminopeptidase A.

Formula: C₁₅H₁₆N₂O₃
Storage Temperature: Ambient
MDL Number: MFCD00037740
CAS Number: 635-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR