BANA

Supplier: BACHEM AMERICAS INC

4000654.0001 4000654.0005
K-1125.0001BAEA 185.3 CAD
K-1125.0001BA K-1125.0005BA
BANA
BANA

Sequence: Bz-DL-Arg-βNA · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Synonym(s): DL-BANA

Formula: C₂₃H₂₆ClN₅O₂
Storage Temperature: Ambient
MDL Number: MFCD00038940
CAS Number: 913-04-2
EINECS: 213-015-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR