H-Asp-βNA

Supplier: BACHEM AMERICAS INC

4002834.0250 4002834.1000
K-1095.0250BAEA 180.91 CAD
K-1095.0250BA K-1095.1000BA
H-Asp-βNA
H-Asp-βNA

Sequence: H-Asp-βNA


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


Substrate for aminopeptidase A.

Formula: C₁₄H₁₄N₂O₃
Storage Temperature: Freezer
MDL Number: MFCD00037742
CAS Number: 635-91-6
EINECS: 211-243-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR