L-Arginine-β-naphthylamide hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Arg-βNA·HCl

4000556.0001 4000556.0005
K-1080.0001BAEA 232.35 CAD
K-1080.0001BA K-1080.0005BA
L-Arginine-β-naphthylamide hydrochloride
L-Arginine-beta-naphthylamide hydrochloride

Sequence: H-Arg-βNA · HCl


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Substrate for aminopeptidase B (arginine aminopeptidase) and cathepsin H.

Formula: C₁₆H₂₂ClN₅O
MW: 335.836 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00051283
CAS Number: 18905-73-2
EINECS: 242-654-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR