Suc-Phe-Leu-Phe-4MbetaNA
Supplier: BACHEM AMERICAS INC
4011631.0050 4011631.0250
J-1320.0050BAEA 1191.53 CAD
J-1320.0050BA J-1320.0250BA
Suc-Phe-Leu-Phe-4MbetaNA
Proteins and Peptides
Suc-FLF-MNA is a substrate for chymotrypsin and Lon proteases as E.coli protease La.