AC1OCUTN
Supplier: BACHEM AMERICAS INC
Synonyms:
Pyr-4MβNA
4031140.0250
4031140.1000
J-1295.0250BAEA
1369.61
CAD
J-1295.0250BA
J-1295.1000BA
AC1OCUTN
AC1OCUTN
Sequence: Pyr-4MβNA
Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.
Glp-MNA
Formula:
C₁₆H₁₆N₂O₃ MW: 284.315 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00237244 CAS Number: 94102-66-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...