H-Pro-4MβNA·HCl

Supplier: BACHEM AMERICAS INC

4000715.0250 4000715.1000
J-1280.0250BAEA 309.81 CAD
J-1280.0250BA J-1280.1000BA
H-Pro-4MβNA·HCl
H-Pro-4MbetaNA·HCl

Sequence: H-Pro-4MβNA · HCl

Formula: C₁₆H₁₉ClN₂O₂
Storage Temperature: Freezer
MDL Number: MFCD00058032
CAS Number: 100930-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR