H-Gly-Pro-4MbetaNA
Supplier: BACHEM AMERICAS INC
4000721.0250 4000721.1000
J-1210.0250BAEA 407.01 CAD
J-1210.0250BA J-1210.1000BA
H-Gly-Pro-4MbetaNA
Proteins and Peptides
See also H-Lys-Pro-AMC (I-1745) and H-Lys-Pro-MNA (J-1255) .
A specific substrate for dipeptidyl aminopeptidase IV (DPP IV). Gly-Pro-MNA is very suitable for histochemical purposes.