H-Glu(4MβNA)-OH
Supplier: BACHEM AMERICAS INC
4001557.0050
4001557.1000
J-1185.0050BAEA
185.46
CAD
J-1185.0050BA
J-1185.1000BA
H-Glu(4MβNA)-OH
H-Glu(4MβNA)-OH
Sequence: H-Glu(4MβNA)-OH
The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.
See also the glutamic acid γ-diazomethyl ketones L-DON (F-1510) and D-DON (F-2185).
Substrate for the determination of γ-glutamyl transferase activity. H-Glu(MNA)-OH is also used in histochemistry.
Formula:
C₁₆H₁₈N₂O₄ Storage Temperature: Freezer |
MDL Number:
MFCD00037728 CAS Number: 24723-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...