H-Arg-4MβNA

Supplier: BACHEM AMERICAS INC

4002530.0250 4002530.1000
J-1035.0250BAEA 413.58 CAD
J-1035.0250BA J-1035.1000BA
H-Arg-4MβNA
H-Arg-4MβNA

Sequence: H-Arg-4MβNA


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also H-Arg(Me)-OH (E-3745) and Fmoc-Arg(Tos)-OH (B-1435).


Substrate for cathepsin. Arg-MNA is especially suitable for the detection of enzyme activity in gel electrophoresis.

Formula: C₁₇H₂₃N₅O₂
Storage Temperature: Freezer
MDL Number: MFCD00058039
CAS Number: 60285-94-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR