Ac-Gly-Pro-AFC
Supplier: BACHEM AMERICAS INC
4053025.0005
4053025.0025
I-2000.0005BAEA
370.51
CAD
I-2000.0005BA
I-2000.0025BA
Ac-Gly-Pro-AFC
Proteins and Peptides
Ac-GP-AFC, specific substrate for the endopeptidase activity of fibroblast activation protein (FAP) (Km = 330 ± 30 μM; kcat = 7.7 ± 0.2 s⁻ ¹).