Bz-Nle-Lys-Arg-Arg-AMC Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4055312.0001
4055312.0005
I-1995.0001BAEA
661.36
CAD
I-1995.0001BA
I-1995.0005BA
Bz-Nle-Lys-Arg-Arg-AMC Trifluoroacetate
Proteins and Peptides
Substrate for dengue virus NS2B-NS3 and yellow fever virus NS3 protease. The kcat/Km value of Bz-NleKRR-AMC for the dengue virus type 4 (DEN4) enzyme is > 800 fold higher than that of Boc-Gly-Arg-Arg-AMC (I-1565), a widely used NS3 substrate (kcat = 2.9 s⁻¹, Km = 8.6 μM for DEN4). For yellow fever virus NS3 protease the kcat value was 0.111 s⁻¹, the Km value 14.6 μM.