Boc-L-Lys-MCA

Supplier: BACHEM AMERICAS INC

4060676.0050 4060676.0250
I-1985.0050BAEA 375.93 CAD
I-1985.0050BA I-1985.0250BA
Boc-L-Lys-MCA
Boc-L-Lys-MCA

Sequence: Boc-Lys(Tfa)-AMC


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.


Fluorogenic substrate for assaying histone deacetylase (HDAC) 4, 5 and 7 activity in a protease-coupled assay. Suitable substrate for HDAC 8. The deacylated product Boc-Lys-AMC corresponds to I-1880.

Formula: C₂₃H₂₈F₃N₃O₆
Storage Temperature: Freezer
MDL Number: MFCD11975034
CAS Number: 97885-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR