Ac-Gly-Ala-Lys(Ac)-AMC
Supplier: BACHEM AMERICAS INC
4060671.0005 4060671.0025
I-1975.0005BAEA 529.25 CAD
I-1975.0005BA I-1975.0025BA
Ac-Gly-Ala-Lys(Ac)-AMC
Proteins and Peptides
See also Boc-Lys(Ac)-AMC (I-1875) and Boc-Lys(Tfa)-AMC (I-1985).
Ac-GAK(Ac)-AMC, fluorogenic substrate for measuring histone deacetylase class I (HDAC 1, 2, 3, and 8) and class II (HDAC 6 and 10) activity in a protease-coupled assay. HDAC-catalyzed deacetylation of Lys yields Ac-GAK-AMC (I-1980).