Ac-Val-Arg-Pro-Arg-AMC Trifluoroacetate

Supplier: BACHEM AMERICAS INC

4048494.0001 4048494.0005
I-1965.0001BAEA 258.14 CAD
I-1965.0001BA I-1965.0005BA
Ac-Val-Arg-Pro-Arg-AMC Trifluoroacetate
Proteins and Peptides

Ac-VRPR-AMC, a fluorogenic substrate for Arabidopsis thaliana metacaspase-9 (AtMC9) (kcat/Km = 4.6 · 10⁵ M⁻¹s⁻¹).

Order Now

SPECIFICATIONS

Protein/peptide name Ac-Val-Arg-Pro-Arg-AMC Trifluoroacetate
Environmentally Preferable
Species
Source
CAS No. 919515-51-8
MDL No. MFCD18782519
Sequence Ac-Val-Arg-Pro-Arg-AMC
Molecular weight 725.85
Molecular formula C₃₄H₅₁N₁₁O₇
Storage conditions –20 ± 5 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR