Z-Leu-Arg-Amc
Supplier: BACHEM AMERICAS INC
4034611.0050
4034611.0250
I-1960.0050BAEA
926.18
CAD
I-1960.0050BA
I-1960.0250BA
Z-Leu-Arg-Amc
Proteins and Peptides
Z-LR-AMC, a fluorogenic substrate for cathepsin K, L, S, and V, human tissue kallikrein, and the falcipains I - III. Z-LR-AMC is the preferred substrate of falcipain II, a hemoglobin-degrading cysteine protease from Plasmodium falciparum, one of the most promising targets for antimalarial drug design.