Z-Leu-Leu-Glu-7-amino-4-methylcoumarin

Supplier: BACHEM AMERICAS INC

Synonyms: Z-Leu-Leu-Glu-AMC

4037381.0005 4037381.0025
I-1945.0005BAEA 651.39 CAD
I-1945.0005BA I-1945.0025BA
Z-Leu-Leu-Glu-7-amino-4-methylcoumarin
Z-Leu-Leu-Glu-7-amino-4-methylcoumarin

Sequence: Z-Leu-Leu-Glu-AMC


Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.


Z-LLE-AMC is an excellent fluorogenic substrate for measuring the peptidylglutamyl-peptide-hydrolyzing (PGPH) activity of the 20S proteasome. See also e.g. I-1085 (for assaying the trypsin-like activity) and I-1395 (for determining the chymotrypsin-like activity).

Formula: C₃₅H₄₄N₄O₉
MW: 664.756 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01074989
CAS Number: 348086-66-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR