Ac-Arg-Gly-Lys(Ac)-AMC

Supplier: BACHEM AMERICAS INC

4044046.0005 4044046.0025
I-1925.0005BAEA 516.34 CAD
I-1925.0005BA I-1925.0025BA
Ac-Arg-Gly-Lys(Ac)-AMC
Ac-Arg-Gly-Lys(Ac)-AMC

Sequence: Ac-Arg-Gly-Lys(Ac)-AMC
Synonym(s): Ac-RGK(Ac)-AMC


See also Boc-Lys(Ac)-AMC (I-1875) and Boc-Lys(Tfa)-AMC (I-1985).


Ac-RGK(Ac)-AMC, fluorogenic substrate for assaying histone deacetylase (HDAC) activity in a two-step enzymatic reaction. The assay consists of the initial lysine deacetylation by HDAC followed by the release of the fluorescent group by trypsin.

Formula: C₂₈H₄₀N₈O₇
Storage Temperature: Freezer
MDL Number: MFCD18782516
CAS Number: 660846-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR