L-Histidine 7-amido-4-methylcoumarin

Supplier: BACHEM AMERICAS INC

Synonyms: H-His-AMC

4015210.0250 4015210.1000
I-1920.0250BAEA 2481.09 CAD
I-1920.0250BA I-1920.1000BA
L-Histidine 7-amido-4-methylcoumarin
L-Histidine 7-amido-4-methylcoumarin

Sequence: H-His-AMC


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₁₆H₁₆N₄O₃
MW: 312.329 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18633529
CAS Number: 191723-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR