H-Met-Gly-Pro-AMC · HCl
Supplier: BACHEM AMERICAS INC
4033312.0025
4033312.0100
I-1900.0025BAEA
1345.09
CAD
I-1900.0025BA
I-1900.0100BA
H-Met-Gly-Pro-AMC · HCl
Proteins and Peptides
MGP-AMC, fluorogenic substrate for methionine aminopeptidases 1D and 2.