Boc-Lys-AMC

Supplier: BACHEM AMERICAS INC

4033973.0250 4033973.1000
I-1880.0250BAEA 903.62 CAD
I-1880.0250BA I-1880.1000BA
Boc-Lys-AMC
Boc-Lys-AMC

Sequence: Boc-Lys-AMC


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
See also Z-Lys-ONp · HCl, M-1295.

Formula: C₂₁H₂₉N₃O₅
Storage Temperature: Freezer
MDL Number: MFCD11041003
CAS Number: 116883-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR