Ac-Nle-Pro-Nle-Asp-AMC
Supplier: BACHEM AMERICAS INC
4030084.0005
4030084.0025
I-1850.0005BAEA
1190.8
CAD
I-1850.0005BA
I-1850.0025BA
Ac-Nle-Pro-Nle-Asp-AMC
Proteins and Peptides
Substrates for measuring the chymotryptic, PGPH, and tryptic acitivity of the 20S or 26S proteasome. See also the thrombin substrate I-1140.
Substrate for proteasome.