Ac-Ile-Glu-Pro-Asp-AMC

Supplier: BACHEM AMERICAS INC

4030484.0005 4030484.0025
I-1835.0005BAEA 529.25 CAD
I-1835.0005BA I-1835.0025BA
Ac-Ile-Glu-Pro-Asp-AMC
Ac-Ile-Glu-Pro-Asp-AMC

Sequence: Ac-Ile-Glu-Pro-Asp-AMC
Synonym(s): Ac-IEPD-AMC


Ac-IEPD-AMC is a preferred substrate sequence of granzyme B, a mammalian serine protease involved in the induction of rapid target cell death by cytotoxic lymphocytes.

Formula: C₃₂H₄₁N₅O₁₁
Storage Temperature: Freezer
MDL Number: MFCD02259622
CAS Number: 216757-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR