Ac-Met-AMC

Supplier: BACHEM AMERICAS INC

4028557.0050 4028557.0250
I-1800.0050BAEA 284.04 CAD
I-1800.0050BA I-1800.0250BA
Ac-Met-AMC
Ac-Met-AMC

Sequence: Ac-Met-AMC


Oxidation of Met to the sulfoxide Met(O) can be easily reverted, whereas sulfone (Met(O₂)) formation is practically irreversible.
See also Boc-Met-ONp, A-1960.


Substrate for acylamino acid-releasing enzyme [EC 3.4.19.1].

Formula: C₁₇H₂₀N₂O₄S
Storage Temperature: Freezer
MDL Number: MFCD02259620
CAS Number: 354152-20-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR