Suc-Ala-Glu-Pro-Phe-AMC
Supplier: BACHEM AMERICAS INC
4027817.0050
4027817.0250
I-1750.0050BAEA
1729.74
CAD
I-1750.0050BA
I-1750.0250BA
Suc-Ala-Glu-Pro-Phe-AMC
Proteins and Peptides
Suc-AEPF-AMC, fluorogenic substrate for the peptidylprolyl isomerase Pin1.