Z-Tyr-Val-Ala-Asp-AFC

Supplier: BACHEM AMERICAS INC

4028224.0005 4028224.0025
I-1720.0005BAEA 317.52 CAD
I-1720.0005BA I-1720.0025BA
Z-Tyr-Val-Ala-Asp-AFC
Z-Tyr-Val-Ala-Asp-AFC

Sequence: Z-Tyr-Val-Ala-Asp-AFC
Synonym(s): Z-YVAD-AFC


The presence of halogen substituents at the fluorescent group improves membrane permeability of the YVAD-derived caspase-1 substrate Z-YVAD-AFC.

Formula: C₃₉H₄₀F₃N₅O₁₁
Storage Temperature: Freezer
MDL Number: MFCD00797534
CAS Number: 201608-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR