Z-Arg-Leu-Arg-Gly-Gly-AMC Acetate
Supplier: BACHEM AMERICAS INC
4027158.0025
4027158.0100
I-1690.0025BAEA
516.34
CAD
I-1690.0025BA
I-1690.0100BA
Z-Arg-Leu-Arg-Gly-Gly-AMC Acetate
Proteins and Peptides
Z-RLRGG-AMC is a fluorogenic substrate for the deubiquitinating enzyme isopeptidase T (IPaseT) and other ubiquitin C-terminal hydrolases (UCHs) based on the C-termini of ubiquitin with a kcat/Km value of 95 M⁻¹s⁻¹. Together with our product I-1685 it is used in continuous assays for precise mechanistic studies or microtiter plate assays for high-throughput inhibitor screening.