L-Tryptophan-7-amido-4-methylcoumarin hydrochloride

Supplier: BACHEM AMERICAS INC

4013433.0250 4013433.1000
I-1670.0250BAEA 1161.77 CAD
I-1670.0250BA I-1670.1000BA
L-Tryptophan-7-amido-4-methylcoumarin hydrochloride
L-Tryptophan-7-amido-4-methylcoumarin hydrochloride

Sequence: H-Trp-AMC

Formula: C₂₁H₂₀ClN₃O₃
Storage Temperature: Freezer
MDL Number: MFCD02683915
CAS Number: 201860-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR