Ac-Asp-Glu-Val-Asp-AMC
Supplier: BACHEM AMERICAS INC
4026262.0005 4026262.0025
I-1660.0005BAEA 585.22 CAD
I-1660.0005BA I-1660.0025BA
Ac-Asp-Glu-Val-Asp-AMC
A continuous fluorometric assay employing Ac-DEVD-AMC has been developed for the measurement of caspase-3, a cysteine protease also called apopain, Yama or CPP-32. The design of this substrate Ac-DEVD-AMC was based on the substrate motif that has been used successfully with caspase-1, using the poly(ADP-ribose) polymerase cleavage site P₁-P₄ tetrapeptide.