Pyr-Arg-Thr-Lys-Arg-AMC Trifluoroacetate
Supplier: BACHEM AMERICAS INC
4018149.0005 4018149.0025
I-1650.0005BAEA 264.68 CAD
I-1650.0005BA I-1650.0025BA
Pyr-Arg-Thr-Lys-Arg-AMC Trifluoroacetate
Proteins and Peptides
The fluorogenic substrate pERTKR-AMC is efficiently cleaved by furin, a mammalian homolog of the yeast Kex2 endoprotease (kcat/Km approx. 2·10⁴ M-¹s-¹). As a calcium-dependent serine protease associated with Golgi membranes, furin is responsible for the secretory processing of precursor proteins at paired basic residues. Also cleaved by proprotein convertase 4. IpERTKR-AMC is an excellent substrate for flavivirin.