Boc-Arg-Val-Arg-Arg-AMC

Supplier: BACHEM AMERICAS INC

4018735.0025 4018735.0100
I-1645.0025BAEA 678.52 CAD
I-1645.0025BA I-1645.0100BA
Boc-Arg-Val-Arg-Arg-AMC
Boc-Arg-Val-Arg-Arg-AMC

Sequence: Boc-Arg-Val-Arg-Arg-AMC


The fluorogenic substrate Boc-RVRR-AMC is efficiently cleaved by furin, a mammalian homolog of the yeast Kex2 endoprotease (kcat/Km approx. 4·10³ M⁻¹s⁻¹). As a calcium-dependent serine protease associated with Golgi membranes, furin is responsible for the secretory processing of precursor proteins at paired basic residues. Boc-RVRR-AMC is also cleaved by proprotein convertase 4. Stock solutions of Boc-RVRR-AMC are best prepared in DMSO.

Formula: C₃₈H₆₂N₁₄O₈
Storage Temperature: Freezer
MDL Number: MFCD02259613
CAS Number: 136132-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR