Ac-Tyr-Val-Ala-Asp-AMC
Supplier: BACHEM AMERICAS INC
4017748.0005
4017748.0025
I-1630.0005BAEA
424.25
CAD
I-1630.0005BA
I-1630.0025BA
Ac-Tyr-Val-Ala-Asp-AMC
Proteins and Peptides
Ac-YVAD-AMC is a fluorogenic substrate for caspase-1 (ICE). It displayed a Km of 8.5 ± 0.5 µM for recombinant human caspase-1.