DTXSID70745476

Supplier: BACHEM AMERICAS INC

Synonyms: Boc-Glu(OBzl)-Ala-Arg-AMC·HCl

4017302.0050 4017302.0250
I-1575.0050BAEA 1429.85 CAD
I-1575.0050BA I-1575.0250BA
DTXSID70745476
DTXSID70745476

Sequence: Boc-Glu(OBzl)-Ala-Arg-AMC · HCl


Boc-EAR-AMC, fluorogenic substrate for coagulation factor XIa and trypsin.

Formula: C₃₆H₄₈ClN₇O₉
Storage Temperature: Freezer
MDL Number: MFCD00133630
CAS Number: 133448-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR