Boc-Asp(OBzl)-Pro-Arg-AMC · HCl
Supplier: BACHEM AMERICAS INC
4017304.0050 4017304.0250
I-1560.0050BAEA 619.6 CAD
I-1560.0050BA I-1560.0250BA
Boc-Asp(OBzl)-Pro-Arg-AMC · HCl
Proteins and Peptides
Highly sensitive substrate for thrombin and trypsin.