Z-Val-Val-Arg-AMC
Supplier: BACHEM AMERICAS INC
4016422.0050
4016422.0250
I-1540.0050BAEA
688.42
CAD
I-1540.0050BA
I-1540.0250BA
Z-Val-Val-Arg-AMC
Z-Val-Val-Arg-AMC
Sequence: Z-Val-Val-Arg-AMC
Z-VVR-AMC is a substrate of human and canine cathepsin S. Z-VVR-AMC is also cleaved by falcipain-related enzymes from Plasmodium species.
Formula:
C₃₄H₄₅N₇O₇ MW: 663.776 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00238523 CAS Number: 124485-41-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...