Suc-Ala-Ala-Pro-Val-AMC
Supplier: BACHEM AMERICAS INC
4015705.0050 4015705.0250
I-1490.0050BAEA 1729.74 CAD
I-1490.0050BA I-1490.0250BA
Suc-Ala-Ala-Pro-Val-AMC
Proteins and Peptides
See also C-1040, C-2345, I-1000, and L-1435.
Suc-AAPV-AMC, substrate for human leukocyte elastase.