Suc-Ala-Ala-Pro-Phe-AMC

Supplier: BACHEM AMERICAS INC

4012873.0050 4012873.0250
I-1465.0050BAEA 793.9 CAD
I-1465.0050BA I-1465.0250BA
Suc-Ala-Ala-Pro-Phe-AMC
Suc-Ala-Ala-Pro-Phe-AMC

Sequence: Suc-Ala-Ala-Pro-Phe-AMC


The peptidylprolyl isomerase substrate Suc-AAPF-AMC is also hydrolyzed by carboxypeptidase Y, cathepsin G, and chymotrypsin. Suc-AAPF-AMC has also been used to analyze the chymotrypsin-like activity of trypsins.

Formula: C₃₄H₃₉N₅O₉
MW: 661.712 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00080246
CAS Number: 88467-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR