L-Citrulline 7-amido-4-methylcoumarin hydrobromide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Cit-AMC·HBr

4019017.0025 4019017.0100
I-1460.0025BAEA 645.43 CAD
I-1460.0025BA I-1460.0100BA
L-Citrulline 7-amido-4-methylcoumarin hydrobromide
L-Citrulline 7-amido-4-methylcoumarin hydrobromide

Sequence: H-Cit-AMC · HBr


Bachem also offers Fmoc-Cit-Wang resin, D-1840.


Synonym(s): H-Orn(carbamoyl)-AMC · HBr

Formula: C₁₆H₂₁BrN₄O₄
Storage Temperature: Freezer
MDL Number: MFCD00152040
CAS Number: 123314-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR